missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylguaiacol 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30317303
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317303 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317303 Supplier Dr. Ehrenstorfer Supplier No. DREA13338000ME1000
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Ethylguaiacol
CAS 2785-89-9
Analyte or Component Names 4-Ethylguaiacol
Shipping Condition Room Temperature
Molecular Formula C9 H12 O2
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Phenol, 4-ethyl-2-methoxy-, Guaiacol, 4-ethyl- (6CI), 4-Ethyl-2-methoxyphenol, 2-Methoxy-4-ethylphenol, 4-Ethylguaiacol, 4-Hydroxy-3-methoxyethylbenzene, 4-Hydroxy-3-methoxyphenylethane, Guaiacyl ethane, NSC 82313, p-Ethylguaiacol
InChI Formula InChI=1S/C9H12O2/c1-3-7-4-5-8(10)9(6-7)11-2/h4-6,10H,3H2,1-2H3
SMILES CCc1ccc(O)c(OC)c1
IUPAC Name 4-ethyl-2-methoxyphenol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 152.19
Quantity 1 mL
Formula Weight 152.0837 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.