missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenoxyacetic acid, 97%, Thermo Scientific™

Product Code. 11304464
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11304464

Brand: Thermo Scientific Alfa Aesar L01102.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24431-27-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00014366
InChI Key WVELHLIHMYYZAT-UHFFFAOYSA-N
Synonym 4-ethylphenoxy acetic acid, 2-4-ethylphenoxy acetic acid, 4-ethylphenoxyacetic acid, 2-p-ethylphenoxy acetic acid, acetic acid, 4-ethylphenoxy, acmc-1cdn7, p-ethylphenoxy acetic acid, cambridge id 5147400, acetic acid,2-4-ethylphenoxy
PubChem CID 90499
IUPAC Name 2-(4-ethylphenoxy)acetic acid
SMILES CCC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 92°C to 94°C
Quantity 5 g
Beilstein 2259506
Formula Weight 180.2
Percent Purity 97%
Chemical Name or Material 4-Ethylphenoxyacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.