missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethylsulfonyl)benzeneboronic acid, 98+%

Product Code. 11304785
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304785 250 mg 250mg
11394775 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304785 Supplier Thermo Scientific Alfa Aesar Supplier No. L17814.MD
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 352530-24-6 | C8H11BO4S | 214.042 g/mol

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352530-24-6
Molecular Formula C8H11BO4S
Molecular Weight (g/mol) 214.042
MDL Number MFCD02093076
InChI Key CVBNDDGEVPNUNA-UHFFFAOYSA-N
Synonym 4-ethylsulfonyl phenylboronic acid, 4-ethylsulfonyl phenyl boronic acid, 4-ethanesulfonyl benzeneboronic acid, 4-ethanesulfonyl phenylboronic acid, 4-ethylsulfonylphenyl boronic acid, 4-ethylsulphonyl benzeneboronic acid, boronic acid, 4-ethylsulfonyl phenyl, 4-ethanesulfonyl phenyl boranediol, 4-ethanesulphonyl benzeneboronic acid
PubChem CID 2773420
IUPAC Name (4-ethylsulfonylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)S(=O)(=O)CC)(O)O

Specifications

Melting Point 135°C to 139°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 214.05
Percent Purity ≥98%
Chemical Name or Material 4-(Ethylsulfonyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.