missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-1,2-dimethoxybenzene, 98%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.
Solubility
Soluble in water 0.079 g/L 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.
Solubility
Soluble in water 0.079 g/L 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Density | 1.171 |
| Boiling Point | 120°C to 123°C (45 mmHg) |
| Flash Point | 91°C (195°F) |
| Refractive Index | 1.51 |
| Quantity | 5 g |
| Beilstein | 2441600 |
| Solubility Information | Soluble in water 0.079g/L 25°C. |
| Formula Weight | 156.16 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Fluoro-1,2-dimethoxybenzene |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction