missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-2-methylanisole, 98%

Product Code. 11428676
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428676 25 g 25g
11418676 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428676 Supplier Alfa Aesar™ Supplier No. A16031.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 399-54-2
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00143319
InChI Key QXOBYWRKNIDHJG-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylanisole, 5-fluoro-2-methoxytoluene, benzene, 4-fluoro-1-methoxy-2-methyl, 4-fluoro-1-methoxy-2-methyl-benzene, pubchem2622, 4-fluoro-2-methylanizole, acmc-1agr3, ksc582c5b, 2-methoxy-5-fluorotoluene
PubChem CID 136239
IUPAC Name 4-fluoro-1-methoxy-2-methylbenzene
SMILES COC1=CC=C(F)C=C1C

Specifications

Boiling Point 69°C to 70°C (4 mmHg)
Refractive Index 1.491
Quantity 25 g
Beilstein 2517140
Formula Weight 140.16
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-methylanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.