missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzeneboronic acid, 98%

Product Code. p-7027398
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11441127

Brand: Thermo Scientific Alfa Aesar B24117.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 139911-29-8 | C7H8BFO2 | 153.947 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139911-29-8
Molecular Formula C7H8BFO2
Molecular Weight (g/mol) 153.947
MDL Number MFCD02093072
InChI Key IQMLIVUHMSIOQP-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp
PubChem CID 2734665
IUPAC Name (4-fluoro-2-methylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)F)C)(O)O

Specifications

Melting Point 199°C to 201°C
Quantity 1 g
Formula Weight 153.95
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-methylbenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.