missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzonitrile, 97%

Product Code. 11312539
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11312539

Brand: Thermo Scientific Alfa Aesar H26641.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 147754-12-9 | C8H6FN | 135.141 g/mol

As organic intermediate, pharmaceutical intermediate. In chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As organic intermediate, pharmaceutical intermediate. In chemical synthesis.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147754-12-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD03095106
InChI Key BJBXUIUJKPOZLV-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorobenzonitrile, 4-fluoro-2-methyl-benzonitrile, benzonitrile, 4-fluoro-2-methyl, 2-cyano-5-fluorotoluene, 4-fluoro-2-methylbenzenecarbonitrile, pubchem1554, intermediates-zcf02129, methyl 4-fluorobenzonitrile, 4-fuoro-2-methylbenzonitrile, ksc174s6l
PubChem CID 3826055
IUPAC Name 4-fluoro-2-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)F)C#N

Specifications

Melting Point 67°C to 71°C
Quantity 25 g
UN Number UN3439
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.