missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzyl alcohol, 99%

Product Code. 15435087
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15435087

Brand: Thermo Scientific Alfa Aesar H32364.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80141-91-9 | C8H9FO | 140.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80141-91-9
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.157
MDL Number MFCD03701058
InChI Key YSULUXOLTMBSFF-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylphenyl methanol, 4-fluoro-2-methylbenzyl alcohol, 4-fluoro-2-methylbenzylalcohol, rarechem al bd 0506, 4-fluoro-2-methylbenzenemethanol, benzenemethanol, 4-fluoro-2-methyl, 4-fluoro-2-methyl-phenyl-methanol, 4-fluoro-2-methylphenyl methan-1-ol, 4'-fluoro-2-methylbenzyl alcohol
PubChem CID 3872017
IUPAC Name (4-fluoro-2-methylphenyl)methanol
SMILES CC1=C(C=CC(=C1)F)CO

Specifications

Quantity 1 g
Formula Weight 140.16
Percent Purity 99%
Chemical Name or Material 4-Fluoro-2-methylbenzyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.