missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenol, 98%, Thermo Scientific™

Product Code. 10370994 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10370994 1 g Glass bottle 1g
10329020 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10370994 Supplier Thermo Scientific Acros Supplier No. 294370010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-72-2
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00075088
InChI Key GKQDDKKGDIVDAG-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
PubChem CID 136295
IUPAC Name 4-fluoro-2-methylphenol
SMILES CC1=C(C=CC(=C1)F)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 38°C
Color Brown to White
Boiling Point 87°C (14.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)OH
Refractive Index 1.5155 to 1.5175
Quantity 1 g
Formula Weight 126.13
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Fluoro-2-methylphenol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-methylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.