missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenol, 98%

Product Code. 11448676
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448676 25 g 25g
11438676 5 g 5g
11458676 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448676 Supplier Thermo Scientific Alfa Aesar Supplier No. A16032.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 452-72-2 | C7H7FO | 126.13 g/mol

4-Fluoro-2-methylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2-methylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-72-2
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00075088
InChI Key GKQDDKKGDIVDAG-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
PubChem CID 136295
IUPAC Name 4-fluoro-2-methylphenol
SMILES CC1=C(C=CC(=C1)F)O

Specifications

Melting Point 32°C to 36°C
Boiling Point 87°C (14 mmHg)
Flash Point 85°C (185°F)
Odor Phenol-like
Refractive Index 1.5165
Quantity 25 g
UN Number UN3261
Beilstein 2041490
Solubility Information Insoluble in water.
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-methylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.