missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenol, 98%

Product Code. p-7023013
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11438676

Brand: Thermo Scientific Alfa Aesar A16032.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 452-72-2 | C7H7FO | 126.13 g/mol

4-Fluoro-2-methylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2-methylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-72-2
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00075088
InChI Key GKQDDKKGDIVDAG-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
PubChem CID 136295
IUPAC Name 4-fluoro-2-methylphenol
SMILES CC1=C(C=CC(=C1)F)O

Specifications

Melting Point 32°C to 36°C
Boiling Point 87°C (14 mmHg)
Flash Point 85°C (185°F)
Odor Phenol-like
Refractive Index 1.5165
Quantity 5 g
UN Number UN3261
Beilstein 2041490
Solubility Information Insoluble in water.
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-methylphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.