missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroanisole, 98%

Product Code. 15497608
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497608 25 g 25g
15487608 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497608 Supplier Thermo Scientific Alfa Aesar Supplier No. H61110.14
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 445-83-0 | C7H6FNO3 | 171.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-83-0
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.13
MDL Number MFCD00013375
InChI Key FWLPYISRFBKEKV-UHFFFAOYSA-N
Synonym 4-fluoro-2-nitroanisole, 4-fluoro-2-nitroanisol, benzene, 4-fluoro-1-methoxy-2-nitro, 4-fluoro-1-methoxy-2-nitro-benzene, 5-fluoro-2-methoxynitrobenzene, 4-fluor-1-methoxy-2-nitrobenzol, pubchem1966, acmc-1amcp, 4-fluoro-2-nitro-anisole, ksc235o8j
PubChem CID 565656
IUPAC Name 4-fluoro-1-methoxy-2-nitrobenzene
SMILES COC1=C(C=C(F)C=C1)[N+]([O-])=O

Specifications

Melting Point 62°C to 64°C
Color Brown-Yellow
Quantity 25 g
Formula Weight 171.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-nitroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.