missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride, Thermo Scientific™

Product Code. 10686280
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10686280

Brand: Thermo Scientific Maybridge MAY00125DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 176225-09-5
Packaging Amber Glass Bottle
Molecular Formula C7H3ClF4O2S
Quantity 5 g
Synonym 4-fluoro-2-trifluoromethyl benzenesulfonyl chloride, 4-fluoro-2-trifluoromethyl benzene-1-sulfonyl chloride, 4-fluoro-2-trifluoromethyl-benzenesulfonyl chloride, 4-fluoro-2-trifluoromethyl benzenesulphonyl chloride, 2-chlorosulphonyl-4-fluorobenzotrifluoride, 4-fluoro-2-trifluoromethyl-benzenesulfonylchloride, 4-fluoro-2-trifluoromethyl benzene sulphonyl chloride, pubchem4276, acmc-1c2ls, 2-chlorosulphonyl-5-fluorobenzotrifluoride
InChI Key IGMYEVQPXWKFQF-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)C(F)(F)F)S(=O)(=O)Cl
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol) 262.603
PubChem CID 2737548
Formula Weight 262.61
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.