missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzyl bromide, 98%, Thermo Scientific™

Product Code. 10301470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10301470 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10301470 Supplier Thermo Scientific Acros Supplier No. 434220010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 206860-48-2
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 141°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H5BrF4
Quantity 1 g
Synonym 4-fluoro-2-trifluoromethyl benzyl bromide, 1-bromomethyl-4-fluoro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-fluoro-2-trifluoromethyl, alpha-bromo-4-fluoro-2-trifluoromethyl toluene, 1-bromomethyl-4-fluoro-2-trifluoromethyl-benzene, pubchem4642, acmc-20ai28, 4-fluoro-2-trifluoromethylbenzyl bromide, 4-fluoro-2-trifluoromethyl-benzyl bromide
InChI Key JMNOONULDANZRZ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)C(F)(F)F)CBr
IUPAC Name 1-(bromomethyl)-4-fluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 257.03
PubChem CID 2737576
Formula Weight 257.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-(trifluoromethyl)benzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.