missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-formyl-benzoic acid, 97%, Thermo Scientific™

Product Code. 10518613
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10518613 250 mg Amber Glass Bottle 250mg
10229602 1 g Amber Glass Bottle 1g
10215854 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10518613 Supplier Thermo Scientific Maybridge Supplier No. MO07323CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-90-7
Molecular Formula C8H5FO3
Molecular Weight (g/mol) 168.123
InChI Key SKPWEADPCJMLID-UHFFFAOYSA-N
Synonym 4-fluoro-3-formyl-benzoic acid, 3-formyl-4-fluorobenzoic acid, 4-fluoro-3-formylbenzoicacid, ksc495c1b
PubChem CID 2794818
IUPAC Name 4-fluoro-3-formylbenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)C=O)F

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 168.12
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-formyl-benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.