missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid, 98%, Alfa Aesar™

Product Code. 15469865
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469865 250 mg 250mg
15459865 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469865 Supplier Thermo Scientific Alfa Aesar Supplier No. H52512.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 874290-69-4
Molecular Formula C9H11BFNO4
Molecular Weight (g/mol) 226.998
MDL Number MFCD08235106
InChI Key WNDRABQWMCWUIO-UHFFFAOYSA-N
Synonym 4-fluoro-3-n-methoxy-n-methyl carbamoyl benzeneboronic acid, 4-fluoro-3-methoxy methyl carbamoyl phenyl boronic acid, 4-fluoro-3-n-methoxy-n-methyl carbamoyl phenylboronic acid, 4-fluoro-3-methoxy methyl carbamoyl phenylboronic acid, 4-fluoro-3-methoxy methyl carbamoyl benzeneboronic acid, n-methoxy-n-methyl 5-borono-2-fluorobenzamide, n-methoxy-n-methyl-5-borono-2-fluorobenzamide
PubChem CID 44717596
IUPAC Name [4-fluoro-3-[methoxy(methyl)carbamoyl]phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)F)C(=O)N(C)OC)(O)O

Specifications

Melting Point 160°C to 163°C
Quantity 250 mg
Formula Weight 227
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.