missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxybenzoic acid, 95%, Thermo Scientific Chemicals

Product Code. 11305495
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11305495 5 g 5g
11395485 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305495 Supplier Thermo Scientific Alfa Aesar Supplier No. L19673.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82846-18-2 | Molecular Formula: C8H7FO3 | Molecular Weight: 170.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82846-18-2
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00272134
InChI Key LWGCZCMLPRMKIZ-UHFFFAOYSA-N
Synonym 3-methoxy-4-fluorobenzoic acid, 4-fluoro-m-anisic acid, benzoic acid, 4-fluoro-3-methoxy, 4-fluoro-3-methoxybenzoicacid, 4-fluoro-3-methoxy-benzoic acid, 4-fluoranyl-3-methoxy-benzoic acid, pubchem1394, acmc-209pqm, ksc495c0r, rarechem al bo 0973
PubChem CID 598436
IUPAC Name 4-fluoro-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)F

Specifications

Melting Point 206°C to 208°C
Quantity 5 g
Beilstein 4178071
Formula Weight 170.14
Percent Purity 95%
Chemical Name or Material 4-Fluoro-3-methoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.