missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific™

Product Code. p-7032065
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438445 250 mg 250mg
15428445 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438445 Supplier Thermo Scientific Alfa Aesar Supplier No. H33394.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373920-75-2
Molecular Formula C10H7F4N
Molecular Weight (g/mol) 217.167
MDL Number MFCD22201073
InChI Key YKXAXCYASRRQID-UHFFFAOYSA-N
Synonym 4-fluoro-3-methyl-5-trifluoromethyl phenylacetonitrile, 2-4-fluoro-3-methyl-5-trifluoromethyl phenyl acetonitrile
PubChem CID 86277680
IUPAC Name 2-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]acetonitrile
SMILES CC1=C(C(=CC(=C1)CC#N)C(F)(F)F)F

Specifica

Quantity 250 mg
UN Number UN3276
Formula Weight 217.16
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetonitrile
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.