missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylaniline, 97%

Product Code. 11462857
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462857 5 g 5g
11472857 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462857 Supplier Thermo Scientific Alfa Aesar Supplier No. A15482.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 452-69-7 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-69-7
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00025294
InChI Key NYMDPDNETOLVBS-UHFFFAOYSA-N
Synonym 2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
PubChem CID 67981
IUPAC Name 4-fluoro-3-methylaniline
SMILES CC1=CC(N)=CC=C1F

Specifications

Melting Point 32°C to 36°C
Density 1.12
Boiling Point 85°C to 86°C (9 mmHg)
Flash Point 95°C (203°F)
Odor Characteristic
Refractive Index 1.537
Quantity 5 g
UN Number UN2811
Beilstein 2935561
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.