missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzyl bromide, 97%, Thermo Scientific™

Product Code. 11372609
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372609 5 g 5g
11362609 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372609 Supplier Thermo Scientific Alfa Aesar Supplier No. H26681.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Pharmaceutical Intermediate. Organic synthesis, tear gas mixture of ortho-, meta-, and para-isomers.

Solubility
Insoluble in water.

Notes
Lachrymatory. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases. Store protected from moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261951-70-6
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD01631425
InChI Key ZZGGOSDHWXCOCA-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylbenzyl bromide, 4-bromomethyl-1-fluoro-2-methylbenzene, 4-fluoro-3-methylbenzylbromide, benzene, 4-bromomethyl-1-fluoro-2-methyl, acmc-1cgif, 4-bromomethyl-1-fluoro-2-methyl-benzene
PubChem CID 2778135
IUPAC Name 4-(bromomethyl)-1-fluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)CBr)F

Specifications

Boiling Point 80°C (3 mmHg)
Refractive Index 1.545
Quantity 5 g
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylbenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.