missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid, 96%

Product Code. 15400386
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400386 250 mg 250mg
15490376 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400386 Supplier Thermo Scientific Alfa Aesar Supplier No. H53101.MD
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 874219-19-9 | C8H9BFNO3 | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874219-19-9
Molecular Formula C8H9BFNO3
Molecular Weight (g/mol) 196.97
MDL Number MFCD08235038
InChI Key DVDSMLPVTUSIFM-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylcarbamoyl phenyl boronic acid, 4-fluoro-3-methylcarbamoyl phenylboronic acid, 4-fluoro-3-methylcarbamoyl benzeneboronic acid, acmc-209qkk, n-methyl 5-borono-2-fluorobenzamide, 4-fluoro-3-methylcarbamoyl phenyl boronicacid, 4-fluoro-3-methyl carbamoyl phenylboronic acid, 4-fluoranyl-3-methylcarbamoyl phenyl boronic acid, boronic acid, 4-fluoro-3-methylamino carbonyl phenyl
PubChem CID 44717580
IUPAC Name [4-fluoro-3-(methylcarbamoyl)phenyl]boronic acid
SMILES CNC(=O)C1=C(F)C=CC(=C1)B(O)O

Specifications

Melting Point 226°C to 228°C
Quantity 250 mg
Formula Weight 196.97
Percent Purity 96%
Chemical Name or Material 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.