missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific™

Product Code. 15471366
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471366 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15471366 Supplier Thermo Scientific Alfa Aesar Supplier No. H54585.AE
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82297-89-0
Molecular Formula C7H6BrFMg
MDL Number MFCD00075303
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 4-fluoro-3-methylphenylmagnesium bromide, olsgdqispracfy-uhfffaoysa-m, bromo 4-fluoro-3-methylphenyl magnesium, 3-methyl-4-fluorophenyl magnesium bromide, 4-fluoro-3-methylphenyl magnesium bromide, 4-fluoro-3-methylphenylmagnesium bromide, 0.50 m in 2-methf, 4-fluoro-3-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 4-fluoro-3-methylphenylmagnesium bromide solution, 1.0 m in thf, grignard reagent
InChI Key OLSGDQISPRACFY-UHFFFAOYSA-M
SMILES CC1=CC([Mg]Br)=CC=C1F
IUPAC Name magnesium;1-fluoro-2-methylbenzene-4-ide;bromide
Molecular Weight (g/mol) 213.33
PubChem CID 2778169
Formula Weight 213.33
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 4-Fluoro-3-methylphenylmagnesium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.