missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid, 97%

Product Code. p-7028748
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15407565

Brand: Thermo Scientific Alfa Aesar H26154.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 878572-17-9 | C8H3F4NO4 | 253.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878572-17-9
Molecular Formula C8H3F4NO4
Molecular Weight (g/mol) 253.109
MDL Number MFCD08282778
InChI Key XCZRYPKVIJEHSU-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitro-5-trifluoromethyl benzoic acid, benzoic acid,4-fluoro-3-nitro-5-trifluoromethyl
PubChem CID 24721083
IUPAC Name 4-fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
SMILES C1=C(C=C(C(=C1C(F)(F)F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 120°C to 124°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 253.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.