missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzenesulfonamide, 97%

Product Code. 15480798
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480798 250 mg 250mg
15470798 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480798 Supplier Thermo Scientific Alfa Aesar Supplier No. H54516.MD
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 406233-31-6 | C6H5FN2O4S | 220.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 406233-31-6
Molecular Formula C6H5FN2O4S
Molecular Weight (g/mol) 220.174
MDL Number MFCD08703185
InChI Key FAYVDRRKPVJSPE-UHFFFAOYSA-N
PubChem CID 16782487
IUPAC Name 4-fluoro-3-nitrobenzenesulfonamide
SMILES C1=CC(=C(C=C1S(=O)(=O)N)[N+](=O)[O-])F

Specifications

Quantity 250 mg
Synonym 4-fluoro-3-nitro-benzenesulfonamide, 4-fluoro-3-nitrobenzene-1-sulfonamide, benzenesulfonamide, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenesulfonamide, 4-fluoro-3-nitrobenzene sulfonamide, 4-f luoro-3-nitrobenzene sulfonamide, benzenesulfonamide,4-fluoro-3-nitro, 4-fluoranyl-3-nitro-benzenesulfonamide
Formula Weight 220.18
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-nitrobenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.