missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzotrifluoride, 96%

Product Code. 10216930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10216930 5 g Glass bottle 5g
10127753 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216930 Supplier Thermo Scientific Acros Supplier No. 200610050
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 367-86-2 | C7H3F4NO2 | 209.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-86-2
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00007059
InChI Key HLDFCCHSOZWKAA-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl-benzene
PubChem CID 67778
IUPAC Name 1-fluoro-2-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Density 1.4900g/mL
Boiling Point 92°C (15.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula FC6H3(NO2)CF3
Refractive Index 1.4610 to 1.463
Quantity 5 g
Beilstein 05, IV, 853
Specific Gravity 1.49
Formula Weight 209.1
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-nitrobenzotrifluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Fluoro-3-nitrobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.