missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride, Thermo Scientific™

Product Code. 10302360
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10302360 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10302360 Supplier Thermo Scientific Maybridge Supplier No. MAY00176DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 1682-10-6
Packaging Amber Glass Bottle
Molecular Formula C7H3ClF4O2S
Quantity 1 g
Synonym 4-fluoro-3-trifluoromethyl benzenesulfonyl chloride, 4-fluoro-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-fluoro-3-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro-3-trifluoromethyl, 5-chlorosulphonyl-2-fluorobenzotrifluoride, 4-fluoro-3-trifluoromethyl benzenesulfonylchloride, 4-fluoro-3-trifluoromethyl-benzenesulfonyl chloride, pubchem8507, acmc-209vib, ksc171m7b
InChI Key AYOKQWBPSUHEQU-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)F
IUPAC Name 4-fluoro-3-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol) 262.603
PubChem CID 2737549
Formula Weight 262.61
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.