missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-alpha-methylstyrene, 95%

Product Code. 11458126
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458126 10 g 10g
11468126 50 g 50g
11478126 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458126 Supplier Thermo Scientific Alfa Aesar Supplier No. A14147.09
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 350-40-3 | C9H9F | 136.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-40-3
Molecular Formula C9H9F
Molecular Weight (g/mol) 136.169
MDL Number MFCD00042297
InChI Key VIXHMBLBLJSGIB-UHFFFAOYSA-N
Synonym 4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene
PubChem CID 67690
IUPAC Name 1-fluoro-4-prop-1-en-2-ylbenzene
SMILES CC(=C)C1=CC=C(C=C1)F

Specifications

Density 1.009
Boiling Point 183°C
Flash Point 53°C (127°F)
Refractive Index 1.511
Quantity 10 g
UN Number UN1993
Beilstein 2039696
Formula Weight 136.17
Percent Purity 95%
Chemical Name or Material 4-Fluoro-alpha-methylstyrene
compliance-icons
Product Identifier
  • 4-Fluoro-alpha-methylstyrene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: 1-Fluoro-4-(isopropenyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.