missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-alpha-toluenesulfonyl chloride, 97%

Product Code. 15404207
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15404207 250 mg 250mg
15494197 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404207 Supplier Thermo Scientific Alfa Aesar Supplier No. H31605.MD
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 103360-04-9 | C7H6ClFO2S | 208.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103360-04-9
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD01631930
InChI Key UUQGWVIRPCRTSA-UHFFFAOYSA-N
Synonym 4-fluorophenyl methanesulfonyl chloride, 4-fluorobenzylsulfonyl chloride, 4-fluoro-phenyl-methanesulfonyl chloride, 4-fluorophenyl methyl sulfonyl chloride, chloro 4-fluorophenyl methyl sulfone, 4-fluorophenyl methanesulphonyl chloride, 4-fluoro-alpha-toluenesulfonyl chloride, benzenemethanesulfonyl chloride, 4-fluoro, 4-fluoro-phenyl-methane sulfonyl chloride, pubchem10056
PubChem CID 2759108
IUPAC Name (4-fluorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)F

Specifications

Melting Point 58°C to 61°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 4-Fluoro-alpha-toluenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.