missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-DL-α-phenylglycine, 98+%, Thermo Scientific™

Product Code. 10139712 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10139712

Brand: Thermo Scientific Acros 367930010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7292-73-1
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.16
MDL Number MFCD00066444,MFCD00042727
InChI Key JKFYKCYQEWQPTM-UHFFFAOYNA-N
Synonym 2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro
PubChem CID 98313
IUPAC Name 2-amino-2-(4-fluorophenyl)acetic acid
SMILES NC(C(O)=O)C1=CC=C(F)C=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 300°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H4CH(NH2)COOH
Quantity 1 g
Formula Weight 169.15
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 4-Fluoro-DL-α-phenylglycine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.