missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroaniline, 98%

Product Code. p-134242 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10677763

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 371-40-4 | C6H6FN | 111.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-40-4
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007829
InChI Key KRZCOLNOCZKSDF-UHFFFAOYSA-N
Synonym p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline
PubChem CID 9731
ChEBI CHEBI:28546
IUPAC Name 4-fluoroaniline
SMILES NC1=CC=C(F)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -1.9°C
Density 1.1570g/mL
Boiling Point 187.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H4NH2
Refractive Index 1.5385 to 1.5405
Quantity 25 mL
Beilstein 12, 597
Merck Index 15, 4200
Specific Gravity 1.157
Solubility Information Solubility in water: 33g/L (20°C). Other solubilities: soluble in acetone,ethanol,xylene alcohol,,carbon tetrachloride and diethyl ether
Formula Weight 111.12
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material 4-Fluoroaniline, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Fluoroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.