missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroaniline, 99%

Product Code. 11457457
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457457 500 g 500g
11437457 25 g 25g
11447457 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457457 Supplier Thermo Scientific Alfa Aesar Supplier No. A15597.36
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 371-40-4 | C6H6FN | 111.12 g/mol

It finds its application as an analytical reagent in the enzymic detection of glucose. It is used as an intermediate in the manufacture of pharmaceuticals and plant growth regulators. It is employed as an intermediate for the production of Flumazenil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an analytical reagent in the enzymic detection of glucose. It is used as an intermediate in the manufacture of pharmaceuticals and plant growth regulators. It is employed as an intermediate for the production of Flumazenil.

Solubility
Slightly soluble in water. Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-40-4
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007829
InChI Key KRZCOLNOCZKSDF-UHFFFAOYSA-N
Synonym p-fluoroaniline, benzenamine, 4-fluoro, 1-amino-4-fluorobenzene, 4-fluorobenzenamine, p-fluorophenylamine, 4-fluoronaniline, aniline, p-fluoro, aniline, 4-fluoro, 4-fluoranilin, para-fluoroaniline
PubChem CID 9731
ChEBI CHEBI:28546
IUPAC Name 4-fluoroaniline
SMILES NC1=CC=C(F)C=C1

Specifications

Melting Point -2°C
Density 1.158
Boiling Point 186°C to 187°C
Flash Point 73°C (163°F)
Odor Recognizable
Refractive Index 1.54
Quantity 500 g
UN Number UN2941
Beilstein 742030
Merck Index 14,4169
Solubility Information Slightly soluble in water. Soluble in chloroform and methanol.
Formula Weight 111.12
Percent Purity 99%
Chemical Name or Material 4-Fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.