missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzeneboronic acid, 97%

Product Code. 11448586
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448586 25 g 25g
11428586 1 g 1g
11438586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448586 Supplier Thermo Scientific Alfa Aesar Supplier No. A15991.14
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 1765-93-1 | C6H6BFO2 | 139.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1765-93-1
Molecular Formula C6H6BFO2
Molecular Weight (g/mol) 139.92
MDL Number MFCD00039136
InChI Key LBUNNMJLXWQQBY-UHFFFAOYSA-N
Synonym 4-fluorobenzeneboronic acid, 4-fluorophenyl boronic acid, 4-fluorophenyl boranediol, p-fluorophenylboronic acid, 4-fluorophenylbornic acid, 4-fluorophenyl dihydroxyboron, 4-fluorophenyl dihydroxyborane, p-fluorophenyl boric acid, 4-fluorophenyl boric acid
PubChem CID 285645
ChEBI CHEBI:48661
IUPAC Name (4-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=C(F)C=C1

Specifications

Melting Point 260°C to 265°C
Quantity 25 g
Beilstein 2829653
Formula Weight 139.92
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzeneboronic acid
compliance-icons
Product Identifier
  • 4-Fluorobenzeneboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Fluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.