missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoic acid, 98%

Product Code. 11418096
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418096 25 g 25g
11428096 100 g 100g
11438096 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418096 Supplier Thermo Scientific Alfa Aesar Supplier No. A14129.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 456-22-4 | C7H5FO2 | 140.113 g/mol

4-Fluorobenzoic acid is used as an intermediate for agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzoic acid is used as an intermediate for agrochemicals and pharmaceuticals.

Solubility
Soluble in alcohol, hot water, methanol, and ether. Very slightly soluble in cold water.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-22-4
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD00002530
InChI Key BBYDXOIZLAWGSL-UHFFFAOYSA-N
Synonym p-fluorobenzoic acid, benzoic acid, 4-fluoro, para-fluorobenzoic acid, 4-fluoro-benzoic acid, benzoic acid, p-fluoro, unii-v5roo2hou4, p-fluoro-benzoic acid, v5roo2hou4, 4-fluorobenzoatd, p-flurobenzoic acid
PubChem CID 9973
ChEBI CHEBI:20364
IUPAC Name 4-fluorobenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)F

Specifications

Melting Point 183°C to 187°C
Density 1.47
Quantity 25 g
Beilstein 1906922
Merck Index 14,4171
Solubility Information Soluble in alcohol,hot water,methanol,and ether. Very slightly soluble in cold water.
Formula Weight 140.11
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzoic acid
compliance-icons
Product Identifier
  • 4-Fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Fluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.