missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile, 97%, Maybridge

Product Code. 10265934
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10265934 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10265934 Supplier Thermo Scientific Maybridge Supplier No. SB01283DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1194-02-1
Assay Percent Range 97%
Molecular Formula C7H4FN
Quantity 1 g
Synonym p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
InChI Key AEKVBBNGWBBYLL-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C#N)F
IUPAC Name 4-fluorobenzonitrile
Molecular Weight (g/mol) 121.114
PubChem CID 14517
Formula Weight 121.11
Chemical Name or Material 4-Fluorobenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.