missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile, 99%

Product Code. 11469953
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469953 5 g 5g
11479953 25 g 25g
11489953 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469953 Supplier Thermo Scientific Alfa Aesar Supplier No. A13028.06
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1194-02-1 | C7H4FN | 121.114 g/mol

4-Fluorobenzonitrile is used as chemical intermediate, solvent for perfumes and pharmaceuticals, stabilizer for chlorinated solvents, HPLC analysis, catalyst and component of transition-metal complex catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzonitrile is used as chemical intermediate, solvent for perfumes and pharmaceuticals, stabilizer for chlorinated solvents, HPLC analysis, catalyst and component of transition-metal complex catalysts.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-02-1
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.114
MDL Number MFCD00001812
InChI Key AEKVBBNGWBBYLL-UHFFFAOYSA-N
Synonym p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
PubChem CID 14517
IUPAC Name 4-fluorobenzonitrile
SMILES C1=CC(=CC=C1C#N)F

Specifications

Melting Point 34°C to 37°C
Boiling Point 186°C to 188°C
Flash Point 65°C (149°F)
Odor Almond-like
Quantity 5 g
Beilstein 2041517
Solubility Information Insoluble in water.
Formula Weight 121.11
Percent Purity 99%
Chemical Name or Material 4-Fluorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.