missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile, 99%

Product Code. 10478350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10478350 5 g Glass bottle 5g
10543141 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10478350 Supplier Thermo Scientific Acros Supplier No. 160810050
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1194-02-1 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-02-1
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001812
InChI Key AEKVBBNGWBBYLL-UHFFFAOYSA-N
Synonym p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
PubChem CID 14517
IUPAC Name 4-fluorobenzonitrile
SMILES C1=CC(=CC=C1C#N)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color White
Boiling Point 188°C (750.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CN
Quantity 5 g
Beilstein 09, I, 138
Formula Weight 121.11
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Fluorobenzonitrile, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.