missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile, 99%

Product Code. 10543141 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10543141

Brand: Thermo Scientific Acros 160810250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1194-02-1 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-02-1
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001812
InChI Key AEKVBBNGWBBYLL-UHFFFAOYSA-N
Synonym p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
PubChem CID 14517
IUPAC Name 4-fluorobenzonitrile
SMILES C1=CC(=CC=C1C#N)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color White
Boiling Point 188°C (750.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CN
Quantity 25 g
Beilstein 09, I, 138
Formula Weight 121.11
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Fluorobenzonitrile, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.