missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzotrifluoride, 98+%

Product Code. 11455363
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455363 25 g 25g
11441013 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455363 Supplier Thermo Scientific Alfa Aesar Supplier No. A13377.14
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 402-44-8 | C7H4F4 | 164.103 g/mol

4-Fluorobenzotrifluoride was used in the synthesis of 2,2?-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2?-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzotrifluoride was used in the synthesis of 2,2′-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2′-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-44-8
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.103
MDL Number MFCD00000399
InChI Key UNNNAIWPDLRVRN-UHFFFAOYSA-N
Synonym 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene
PubChem CID 67873
IUPAC Name 1-fluoro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)F

Specifications

Melting Point -42°C
Density 1.295
Boiling Point 103°C to 105°C
Flash Point 10°C (50°F)
Refractive Index 1.401
Quantity 25 g
UN Number UN1993
Beilstein 1909390
Solubility Information Slightly soluble in water.
Formula Weight 164.1
Percent Purity ≥98%
Chemical Name or Material 4-Fluorobenzotrifluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.