missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoyl chloride, 98%

Product Code. 11489783
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489783 500 g 500g
11469783 25 g 25g
11479783 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489783 Supplier Thermo Scientific Alfa Aesar Supplier No. A12946.36
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 403-43-0 | C7H4ClFO | 158.556 g/mol

It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of polyphenylene ether and thioether ketones and also in the synthesis fluorinated poly(aryl ether ketone)s containing 1,4-naphthalene moieties. It is also used as a raw material in engineering of plastics.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-43-0
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000684
InChI Key CZKLEJHVLCMVQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride
PubChem CID 67879
IUPAC Name 4-fluorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)F

Specifications

Melting Point 10°C to 12°C
Density 1.326
Boiling Point 193°C
Flash Point 82°C (179°F)
Odor Acidic
Refractive Index 1.532
Quantity 500 g
UN Number UN3265
Beilstein 386215
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 158.56
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.