missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl chloride, 98%

Product Code. 10685342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10685342 25 mL Glass bottle 25mL
10570921 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10685342 Supplier Thermo Scientific Acros Supplier No. 119430250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 352-11-4 | C7H6ClF | 144.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-11-4
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.58
MDL Number MFCD00000913
InChI Key IZXWCDITFDNEBY-UHFFFAOYSA-N
Synonym 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene
PubChem CID 9602
IUPAC Name 1-(chloromethyl)-4-fluorobenzene
SMILES C1=CC(=CC=C1CCl)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -18°C
Color Yellow
Density 1.2070g/mL
Boiling Point 181°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2Cl
Refractive Index 1.5120 to 1.514
Quantity 25 mL
Specific Gravity 1.207
Formula Weight 144.58
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4-Fluorobenzyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.