missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl chloride, 99%

Product Code. 11477676
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477676 500 g 500g
11457676 25 g 25g
11467676 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477676 Supplier Thermo Scientific Alfa Aesar Supplier No. A14686.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 352-11-4 | C7H6ClF | 144.573 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-11-4
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000913
InChI Key IZXWCDITFDNEBY-UHFFFAOYSA-N
Synonym 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene
PubChem CID 9602
IUPAC Name 1-(chloromethyl)-4-fluorobenzene
SMILES C1=CC(=CC=C1CCl)F

Specifications

Melting Point -18°C
Density 1.214
Boiling Point 185°C
Flash Point 69°C (156°F)
Odor Pungent
Refractive Index 1.513
Quantity 500 g
UN Number UN3265
Beilstein 742272
Formula Weight 144.58
Percent Purity 99%
Chemical Name or Material 4-Fluorobenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.