missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl isocyanate, 98%

Product Code. 10301534 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10301534

Brand: Thermo Scientific Acros 319300050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132740-43-3 | C8H6FNO | 151.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132740-43-3
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00673062
InChI Key HHSIWJYERNCLKQ-UHFFFAOYSA-N
Synonym 4-fluorobenzyl isocyanate, 1-fluoro-4-isocyanatomethyl benzene, 4-fluorobenzylisocyanate, 4-fluorophenyl methanisocyanate, 4-fluorobezylisocyanate, acmc-1c0to, intermediates-zcf02017, 4-fluoro benzyl isocyanate, 4-fluorophenyl methyl isocyanate
PubChem CID 2733376
IUPAC Name 1-fluoro-4-(isocyanatomethyl)benzene
SMILES FC1=CC=C(CN=C=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1860g/mL
Boiling Point 211.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2NCO
Refractive Index 1.5040 to 1.5060
Quantity 5 g
Specific Gravity 1.186
Formula Weight 151.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.