missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl mercaptan, 96%, Thermo Scientific™

Product Code. 11617968 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11617968 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11617968 Supplier Thermo Scientific Acros Supplier No. 443900010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 15894-04-9
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1570g/mL
Boiling Point 72°C to 74°C (15.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Molecular Formula C7H7FS
Quantity 1 g
Specific Gravity 1.157
Synonym 4-fluorobenzyl mercaptan, 4-fluorophenyl methanethiol, 4-fluorobenzylmercaptan, 4-fluoro benzyl mercaptan, 4-fluorobenzenemethanethiol, p-fluorotoluene-alpha-thiol, benzenemethanethiol, 4-fluoro, 4-fluoro-phenyl-methanethiol, 4-fluorobenzylthiol, 4-fluorobenzyl thiol
InChI Key RKTRHMNWVZRZJQ-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CS)F
IUPAC Name (4-fluorophenyl)methanethiol
Molecular Weight (g/mol) 142.2
PubChem CID 85171
Formula Weight 142.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzyl mercaptan
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzyl mercaptan
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.