missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzylamine, 98+%

Product Code. 11412867
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412867 10 g 10g
11422867 50 g 50g
11432867 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412867 Supplier Thermo Scientific Alfa Aesar Supplier No. A15486.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 140-75-0 | C7H8FN | 125.146 g/mol

4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agent, air, carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-75-0
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00008120
InChI Key IIFVWLUQBAIPMJ-UHFFFAOYSA-N
Synonym 4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392
PubChem CID 67326
IUPAC Name (4-fluorophenyl)methanamine
SMILES C1=CC(=CC=C1CN)F

Specifications

Density 1.095
Boiling Point 182°C to 183°C
Flash Point 66°C (150°F)
Odor Amine-like
Refractive Index 1.513
Quantity 10 g
UN Number UN2735
Beilstein 636501
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 125.15
Percent Purity ≥98%
Chemical Name or Material 4-Fluorobenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.