missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzylamine, 98+%

Product Code. 11422867
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11422867 50 g 50g
11412867 10 g 10g
11432867 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11422867

Brand: Thermo Scientific Alfa Aesar A15486.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-75-0 | C7H8FN | 125.146 g/mol

4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agent, air, carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-75-0
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00008120
InChI Key IIFVWLUQBAIPMJ-UHFFFAOYSA-N
Synonym 4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392
PubChem CID 67326
IUPAC Name (4-fluorophenyl)methanamine
SMILES C1=CC(=CC=C1CN)F

Specifications

Density 1.095
Boiling Point 182°C to 183°C
Flash Point 66°C (150°F)
Odor Amine-like
Refractive Index 1.513
Quantity 50 g
UN Number UN2735
Beilstein 636501
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 125.15
Percent Purity ≥98%
Chemical Name or Material 4-Fluorobenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Fluorobenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.