missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Fluorobenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Specifications
Specifications
| CAS | 312693-07-5 |
| Molecular Formula | C7H6ClFZn |
| MDL Number | MFCD01319879 |
| Quantity | 50 mL |
| UN Number | UN3399 |
| Synonym | zinc 2+ ion 4-fluorophenyl methanide chloride |
| InChI Key | HEXMIUITECMJJV-UHFFFAOYSA-M |
| SMILES | [CH2-]C1=CC=C(C=C1)F.Cl[Zn+] |
| IUPAC Name | chlorozinc(1+);1-fluoro-4-methanidylbenzene |
| Molecular Weight (g/mol) | 209.953 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction