missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorocinnamic acid, 98+%

Product Code. 11442708
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442708 100 g 100g
11422708 5 g 5g
11432708 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442708 Supplier Thermo Scientific Alfa Aesar Supplier No. A16707.22
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 459-32-5 | C9H7FO2 | 166.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-32-5
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.151
MDL Number MFCD00004395
InChI Key ISMMYAZSUSYVQG-ZZXKWVIFSA-N
Synonym 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e
PubChem CID 1530234
IUPAC Name (E)-3-(4-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)F

Specifications

Melting Point 208°C to 210°C
Odor Odorless
Quantity 100 g
Beilstein 2613441
Formula Weight 166.15
Percent Purity ≥98%
Chemical Name or Material 4-Fluorocinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.