missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorocinnamic acid, 99%

Product Code. 10646442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10646442 5 g Glass bottle 5g
10656442 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10646442 Supplier Thermo Scientific Acros Supplier No. 207110050
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 459-32-5 | C9H7FO2 | 166.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-32-5
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.15
MDL Number MFCD00004395
InChI Key ISMMYAZSUSYVQG-ZZXKWVIFSA-N
Synonym 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e
PubChem CID 1530234
IUPAC Name (E)-3-(4-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 207.0°C to 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H4CH=CHCO2H
Quantity 5 g
Beilstein 09, II, 395
Formula Weight 166.15
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Fluorocinnamic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorocinnamic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.