missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroindole, 97%

Product Code. 15482428
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482428 250 mg 250mg
15462428 1 g 1g
15472428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482428 Supplier Thermo Scientific Alfa Aesar Supplier No. H56011.MD
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 387-43-9 | C8H6FN | 135.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 387-43-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00055992
InChI Key ZWKIJOPJWWZLDI-UHFFFAOYSA-N
Synonym 4-fluoroindole, 1h-indole, 4-fluoro, 4-fluoro-indole, pubchem7297, acmc-1cnb8, ksc224m5j, bidd:gt0103
PubChem CID 2774502
IUPAC Name 4-fluoro-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)F

Specifications

Melting Point 30°C to 32°C
Boiling Point 90°C (0.4 mmHg)
Quantity 250 mg
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 4-Fluoroindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.