missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoromandelic acid, 98%

Product Code. 11415393
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415393 1 g 1g
11425393 5 g 5g
11435393 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415393 Supplier Thermo Scientific Alfa Aesar Supplier No. A13391.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 395-33-5 | C8H7FO3 | 170.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 395-33-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD00044402,MFCD20638902
InChI Key RWCMOQXHIDWDDJ-UHFFFAOYNA-N
Synonym 4-fluoromandelic acid, 2-4-fluorophenyl-2-hydroxyacetic acid, p-fluoro mandelic acid, 4-fluorophenyl hydroxy acetic acid, p-fluoromandelic acid, 4-fluoro mandelic acid, 4-fluoromandelicacid, 4-fluoro-alpha-hydroxybenzeneacetic acid, 4-fluoro-dl-mandelic acid, 4-fluoro hydroxy acetic acid
PubChem CID 98068
IUPAC Name 2-(4-fluorophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=C(F)C=C1

Specifications

Melting Point 133°C to 138°C
Quantity 1 g
Beilstein 2211001
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material 4-Fluoromandelic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.